Få mere at vide
Caffeine-13C, TRC
Artikelnummer. 30005131
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005131 100 mg
30004089 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005131 Leverandør Toronto Research Chemicals Leverandørnr. TRCC080103100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202282-98-2
Molekylær formel C713CH10N4O2
Molekylvægt (g/mol) 195.18
Synonym 3,7-Dihydro-1,7-dimethyl-3-(methyl-13C)-1H-Purine-2,6-dione
IUPAC navn 1,7-dimethyl-3-(113C)methylpurine-2,6-dione
SMIL O=C(N1C)C(N2C)=C(N=C2)N([13CH3])C1=O
Tekniske data
Kemisk navn eller materiale Caffeine-13C
Mængde 100 mg
InChI formel InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i2+1