Få mere at vide
Cafedrine Hydrochloride, LoGiCal
Artikelnummer. 30461753
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30461753 5 mg
1 options
Artikelnummer. 30461753 Leverandør LoGiCal Leverandørnr. LGCFOR1180.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cafedrine Hydrochloride
CAS 3039-97-2
Analyt- eller komponentnavne Cafedrine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C18 H23 N5 O3 . Cl H
Certificeringer/compliance ISO 9001
Synonym 1H-Purine-2,6-dione, 3,7-dihydro-7-[2-[[(1S,2R)-2-hydroxy-1-methyl-2-phenylethyl]amino]ethyl]-1,3-dimethyl-, monohydrochloride (9CI), 1H-Purine-2,6-dione, 3,7-dihydro-7-[2-[(2-hydroxy-1-methyl-2-phenylethyl)amino]ethyl]-1,3-dimethyl-, monohydrochloride, [R-(R*,S*)]-, Theophylline, 7-[2-[(β-hydroxy-α-methylphenethyl)amino]ethyl]-, monohydrochloride (8CI), Norephedrine theophylline, [R-(R*,S*)]-3,7-Dihydro-7-[2-[(2-hydroxy-1-methyl-2-phenylethyl)amino]ethyl]-1,3-dimethyl-1H-purine-2,6-dione monohydrochloride, Cafedrine Hydrochloride
InChI formel InChI=1S/C18H23N5O3.ClH/c1-12(15(24)13-7-5-4-6-8-13)19-9-10-23-11-20-16-14(23)17(25)22(3)18(26)21(16)2;/h4-8,11-12,15,19,24H,9-10H2,1-3H3;1H/t12-,15-;/m0./s1
Løsningstype Neat
SMIL Cl.C[C@H](NCCn1cnc2N(C)C(=O)N(C)C(=O)c12)[C@H](O)c3ccccc3
IUPAC navn 7-[2-[[(1R,2S)-1-hydroxy-1-phenylpropan-2-yl]amino]ethyl]-1,3-dimethylpurine-2,6-dione;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 393.87
Mængde 5 mg
Formel vægt 393.1568 g/mol