Learn More
Alfa Aesar™ Caerulein, Sulfated
For cellular and molecular biology applications
Brand: Alfa Aesar™ J64320.|D
Additional Details : CAS Number : 17650-98-5 Weight : 0.00001kg
Specifications
1352.40 | |
0.5mg | |
Powder | |
RUO | |
5422487 | |
Caerulein, sulfated |
HPLC | |
Stimulates smooth muscle and increases digestive secretions | |
Glp-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2 | |
-30° to -10°C | |
Ceruletide | |
Powder |
For Research Use Only