Få mere at vide
Tocris Bioscience™ Caerulein
GREENER_CHOICE
CCK agonist
Brand:  Tocris Bioscience™ 6264/1
Encompass
Produktkode 15746835
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Caerulein is a CCK agonist. Stimulates biosynthesis of pancreatic enzymes in AR4-2J pancreatic cells in vitro. Induces acute pancreatitis in mice. Also enhances proteasome activity in Gdeg mice.
Tekniske data
Tekniske data
Human
Store at −20°C
1 mg
XQDYTGWMDF
17650-98-5
1352.4g/mol
RUO - research use only
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only