Få mere at vide
Cadherin-4/R-Cadherin Antibody, Novus Biologicals™
Artikelnummer. 18648115 Shop alle Bio Techne produkter
Skift visning
Mængde:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18648115 100 μg
18668305 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18648115 Leverandør Novus Biologicals Leverandørnr. NBP299097100ul
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
Cadherin-4/R-Cadherin Polyclonal antibody specifically detects Cadherin-4/R-Cadherin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen Cadherin-4/R-Cadherin
Applikationer ELISA
Klassifikation Polyclonal
Konjugeret Unconjugated
Fortynding ELISA 1:5000-1:10000
Formulering 0.2 μm filtered solution in PBS
Gene Alias CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin
Værtsarter Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Cadherin-4/R-Cadherin (Accession#: NP_001785.2; Met1-Ala734).
Oprensningsmetode Antigen and protein A Affinity-purified
Mængde 100 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 1002
Målarter Human
Indhold og opbevaring Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG