Learn More
Cadence Science™ Reusable Small Animal Feeding Needles: Straight
Product Code. 11737614
Change view
Click to view available options
Diameter (English) Ball:
0.088 in.
Gauge:
18G
20G
Length (English):
1 in.
1.5 in.
2 in.
Unit Size:
Pack of 12
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Ball Gauge Length (English) Unit Size
11737614 0.088 in. 18G 1.5 in. Pack of 12
11747614 0.088 in. 18G 1 in. Pack of 12
17851964 0.088 in. 20G 1 in. Pack of 12
15350053 0.088 in. 18G 2 in. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11737614 Supplier Cadence Science™ Supplier No. 7904
Forventet forsendelse: 23-12-2026
Precision-assembled stainless-steel feeding needles ideal for feeding and dosing
  • For the intubation of laboratory animals
  • Straight, curved tubing styles
  • Many standard sizes available, from small to large
  • Straight
  • Autoclavable
Small Animal Needles
  • For animals 0.7 oz. to 1 lb., 5 oz. (20 to 600g)
Specifications
Autoclavable Autoclavable
Product Type Animal Feeding Needle (Straight)
Material Stainless Steel
Disposable Reusable
For Use With (Application) Small animal, Ideal for feeding and dosing
Gauge 18G
Length (English) 1.5 in.
Length (Metric) 0.38 cm
Diameter (English) Ball 0.088 in.
Diameter (Metric) Ball 0.023 cm