Learn More
Cadence Science™ Laboratory Pipetting Needles
Click to view available options
Needle Gauge:
14 G
16 G
18 G
20 G
22 G
24 G
Length (English) Needle:
1 in.
1.5 in.
2 in.
4 in.
6 in.
Material:
Nickel-plated (Hub), Stainless Steel
Stainless Steel, Stainless Steel (Hub)
Connection Type:
Female Luer
Luer
Unit Size:
Pack of 12
Description
316 Stainless Steel Hub with 316 Stainless Steel Tubing
- Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
- Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate™ Threaded Female Luer Hubs and 316 stainless steel cannulae
- All have 90° blunt ends
Specifications
Specifications
Material | Nickel-plated (Hub), Stainless Steel |
Needle Gauge | 18 G |
For Use With (Application) | For both manual and automatic volumetric dispensers, diluters, pipetters and for laboratory procedures |
Length (English) Needle | 4 in. |
Product Type | Pipetting Needle |
Quantity | 12 Needles |
Needle Point Style | 90° Blunt |
Connection Type | Luer |
Product Content Correction
Product Title
Cadence Science™ Laboratory Pipetting Needles >
Spot an opportunity for improvement?Share a Content Correction