Få mere at vide
Invitrogen™ CACNG8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114707
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Antibody detects endogenous levels of total CACNG8.
Regulates the trafficking and gating properties of AMPA- selective glutamate receptors (AMPARs). Promotes their targeting to the cell membrane and synapses and modulates their gating properties by slowing their rates of activation, deactivation and desensitization and by mediating their resensitization. Does not show subunit-specific AMPA receptor regulation and regulates all AMPAR subunits. Thought to stabilize the calcium channel in an inactivated (closed) state.
Tekniske data
| CACNG8 | |
| Polyclonal | |
| Unconjugated | |
| CACNG8 | |
| CACNG6; Cacng8; calcium channel, voltage-dependent, gamma subunit 8; calcium voltage-gated channel auxiliary subunit gamma 8; CaVGamma8; mir-935; Neuronal voltage-gated calcium channel gamma-8 subunit; TARP gamma 8; TARP gamma-8; Transmembrane AMPAR regulatory protein gamma-8; voltage-dependent calcium channel gamma-8 subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140729, 59283, 81905 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8VHW2, Q8VHW5, Q8WXS5 | |
| CACNG8 | |
| A synthesized peptide derived from human CACNG8(Accession Q8WXS5), corresponding to amino acid residues V167-E217. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |