missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CABP5 Antibody (016), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90251R
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CABP5 Monoclonal specifically detects CABP5 in Human samples. It is validated for ELISA.
Tekniske data
| CABP5 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 56344 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 16 | |
| ELISA | |
| CABP3, calcium binding protein 5, calcium-binding protein 3, calcium-binding protein 5 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CABP5 (Q9NP86; Met1-Arg173). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion