Få mere at vide
Cabergoline-d5, TRC
Artikelnummer. 30334270
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30334270 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334270 Leverandør Toronto Research Chemicals Leverandørnr. TRCC0500020.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cabergoline-d5
CAS 1426173-20-7
Molekylær formel C26H32D5N5O2
Mængde 0.25 mg
Synonym (8β)-N-[3-(Dimethylamino)propyl]-N-[(ethylamino)carbonyl]-6-(2-propen-1-yl)-ergoline-8-carboxamide-d5, FCE-21336-d5, Cabaser-d5, Dostinex-d5
InChI formel InChI=1S/C26H37N5O2/c1-5-11-30-17-19(25(32)31(26(33)27-6-2)13-8-12-29(3)4)14-21-20-9-7-10-22-24(20)18(16-28-22)15-23(21)30/h5,7,9-10,16,19,21,23,28H,1,6,8,11-15,17H2,2-4H3,(H,27,33)
SMIL [H][C@]12C[C@H](C(N(C(NCC)=O)CCCN(C)C)=O)CN(C([2H])([2H])/C([2H])=C([2H])/[2H])[C@]1([H])CC3=CNC4=CC=CC2=C34
IUPAC navn N-[3-(dimethylamino)propyl]-N-(ethylcarbamoyl)-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
Molekylvægt (g/mol) 456.64