Få mere at vide
Cabazitaxel-d6, TRC
Artikelnummer. 30339758
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30339758 10 mg
30334453 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339758 Leverandør Toronto Research Chemicals Leverandørnr. TRCC04650210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cabazitaxel-d6
CAS 1383561-29-2
Molekylær formel C45H51D6NO14
Mængde 10 mg
Synonym (αR,βS)-β-[[(1,1-Dimethylethoxy)carbonyl]amino]-α-hydroxybenzenepropanoic Acid (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(Acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4,6-(dimethoxy-d6)-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl Ester, TXD 258-d6, XRP 6258-d6, β-[[(1,1-Dimethylethoxy)carbonyl]amino]-α-hydroxybenzenepropanoic Acid [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-12b-(Acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4,6-(dimethoxy-d6)-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl Ester
InChI formel InChI=1S/C45H57NO14/c1-24-28(57-39(51)33(48)32(26-17-13-11-14-18-26)46-40(52)60-41(3,4)5)22-45(53)37(58-38(50)27-19-15-12-16-20-27)35-43(8,36(49)34(55-10)31(24)42(45,6)7)29(54-9)21-30-44(35,23-56-30)59-25(2)47/h11-20,28-30,32-35,37,48,53H,21-23H2,1-10H3,(H,46,52)/t28-,29?,30+,32-,33+,34?,35-,37-,43?,44-,45+/m0/s1/i9D3,10D3
SMIL O[C@@]1([C@@H](OC(C2=CC=CC=C2)=O)[C@@]([C@@]3(OC(C)=O)[C@H](OC3)C[C@@H]4OC([2H])([2H])[2H])([H])[C@]4(C)C([C@@H]5OC([2H])([2H])[2H])=O)C[C@H](OC([C@H](O)[C@H](C6=CC=CC=C6)NC(OC(C)(C)C)=O)=O)C(C)=C5C1(C)C
IUPAC navn (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-acetoxy-9-(((2R,3S)-3-((tert-butoxycarbonyl)amino)-2-hydroxy-3-phenylpropanoyl)oxy)-11-hydroxy-4,6-bis(methoxy-d3)-4a,8,13,13-tetramethyl-5-oxo-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-1H-7,11-methanocyclodeca[3,4]benzo[1,2-b]oxet-12-yl benzoate
Molekylvægt (g/mol) 841.97
Formel vægt 841.42