Få mere at vide
Tocris Bioscience™ C26-A6
Beskrivelse
C26-A2 inhibits the interaction between MTDH and SND1 by blocking the SND1 pocket. C26-A2 disrupts MTDH-SND1 complex-induced reduction in tumor antigen presentation and T cell infiltration; enhancing immune surveillance and sensitivity to anti-PD-1 therapy in metastatic breast cancer mouse models. C26-A6 suppresses tumor growth (IC50 = 12.3 μM) and metastasis and enhances chemotherapy sensitivity in triple-negative breast cancer (TNBC) mouse models.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 5-Chloro-2-methoxy-N-(2-methyl[1,2,4 ]triazolo[1,5-a]pyridin-8-yl)benzenesulfonamide |
| CAS | 1341030-00-9 |
| Mængde | 10 mg |
| Mål | Cell Adhesion Inhibitors |
| Molekylær formel | C14H13ClN4O3S |
| Renhed | 0.98 |