missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
C1orf174 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-70453
Denne vare kan ikke returneres.
Se returpolitik
Description
C1orf174 Polyclonal specifically detects C1orf174 in Human samples. It is validated for Western Blot.
Spécification
| C1orf174 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C1ORF174 | |
| Synthetic peptides corresponding to C1ORF174 The peptide sequence was selected from the middle region of C1ORF174. Peptide sequence EAGVSVQQGAASLPLGGCRVVSDSRLAKTRDGLSVPKHSAGSGAEESNSS. | |
| Affinity purified | |
| RUO | |
| 339448 | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| chromosome 1 open reading frame 174, RP13-531C17.2 | |
| Rabbit | |
| 26 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu