missing translation for 'onlineSavingsMsg'
Få mere at vide

C14orf119 Rabbit anti-Human, Polyclonal, Proteintech

Artikelnummer. 16814675 Shop alle Proteintech produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
20 μL
150 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16814675 150 μL
16824675 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16814675 Leverandør Proteintech Leverandørnr. 261411AP150UL

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen C14orf119
Applikationer Western Blot
Klassifikation Polyclonal
Koncentration 0.24 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene C14orf119
Genadgangsnr. Q9NWQ9
Gene Alias C14orf119
Gensymboler C14orf119
Værtsarter Rabbit
Immunogen C14orf119 Fusion Protein Ag23415
Oprensningsmetode Antigen Affinity Chromatography
Mængde 150 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 55017
Målarter Human
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.