Beskrivelse
The function of this protein remains unknown.
Tekniske data
Tekniske data
| Antigen | C14orf119 |
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Koncentration | 0.24 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | C14orf119 |
| Genadgangsnr. | Q9NWQ9 |
| Gene Alias | C14orf119 |
| Gensymboler | C14orf119 |
| Vis mere |