Få mere at vide
C10-C16 Alkyl Ethoxylate Sulfuric Acid Sodium Salt, TRC
Artikelnummer. 30416780
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30416780 1 g
1 options
Artikelnummer. 30416780 Leverandør Toronto Research Chemicals Leverandørnr. TRCS2247201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale C10-C16 Alkyl Ethoxylate Sulfuric Acid Sodium Salt
CAS 68585-34-2
Mængde 1 g
Synonym Alcohol Ethoxysulfate (Sodium Salt)