missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ C-telopeptide
For cellular and molecular biology applications
Brand: Alfa Aesar™ J66595.|D
Additional Details : Weight : 0.00001kg
Specifications
868.89 | |
0.5mg | |
Glu-Lys-Ala-His-Asp-Gly-Gly-Arg | |
-30° to -10°C | |
C-telopeptide |
HPLC | |
Solid | |
RUO | |
C-terminal telopeptide | |
Solid |
For Research Use Only