missing translation for 'onlineSavingsMsg'
Learn More

c-Kit Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA516458

Additional Details : Weight : 0.01000kg

Product Code. 11335783

  • 3160.00 DKK / 500µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PA5-16458 targets CD117 in IHC (P) applications and shows reactivity with Human samples. The PA5-16458 immunogen is a synthetic peptide from C-terminus of human CD117/c-kit protein.

c-Kit Polyclonal antibody specifically detects c-Kit in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

c-Kit
Polyclonal
Unconjugated
KIT
belly-spot; Bs; CD117; ckit; C-Kit; c-KIT gene1; c-kit protein; c-kit proto-oncogene protein; c-kit receptor; c-kit receptor tyrosine kinase; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; kit oncogene; KIT proto-oncogene receptor tyrosine kinase; KIT proto-oncogene, receptor tyrosine kinase; mast cell growth factor receptor; mast/stem cell growth factor receptor; mast/stem cell growth factor receptor Kit; MGF; p145 c-kit; PBT; Piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; receptor tyrosine kinase type III; SCFR; SCO1; SCO5; Sl; soluble KIT variant 1; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; tyrosine kinase receptor; tyrosine kinase receptor c-KIT; tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein; W
Rabbit
Protein A
RUO
3815
4° C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P10721
KIT
A synthetic peptide from C-terminus of human CD117/c-kit protein
500 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers