Få mere at vide
Ekastu Safety™ Eye Wash Bottle MINI with Funnel
Beskrivelse
- Eye Wash Bottle with funnel
- DIN EN 15154-4
- Supplied empty or filled
- For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
- PE bottle with screwed-on eye funnel and tilting valve
- Can be used in both upright and horizontal positions
- leak-proof
- Stores easily in EPS Eye Wash Station (sold separately)
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 200 mL |
| Certificeringer/compliance | EN 15154-4 DIN |
| Inkluderer | Saltvand ikke inkluderet |
| Materiale | Polyethylene |
| Produkttype | Eye Wash Bottle |