missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ C/EBP zeta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599598
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Antibody detects endogenous levels of total C/EBP zeta.
This gene belongs to the CEBP family. The encoded protein plays a role in cellular response to environmental stimuli through a transcriptional process that involves heat shock factors, conserved DNA elements (heat shock elements or HSEs) and CCAAT boxes. The protein acts as a DNA-binding transcriptional activator and regulates the heat-shock protein 70 (HSP70) promoter in a CCAAT-dependent manner. The protein is also involved in cell growth and differentiation, particularly, hematopoietic differentiation. Methylation of the promoter of this gene or mutations within the gene may be correlated with occurrence of acute myeloid leukemia (AML).
Tekniske data
| C/EBP zeta | |
| Polyclonal | |
| Unconjugated | |
| CEBPZ | |
| AI848081; C EBP; C/EBP zeta; CBF; Cbf2; CCAAT/enhancer binding protein (C/EBP), zeta; CCAAT/enhancer binding protein alpha (C/EBP) related sequence 1; CCAAT/enhancer binding protein cebp family zeta; CCAAT/enhancer binding protein zeta; CCAAT/enhancer-binding protein zeta; CCAAT-binding factor; CCAAT-box-binding transcription factor; CEBP zeta; Cebpa-rs1; CEBPZ; core-binding factor; CTF2; HSPCBF; HSP-CBF; NOC1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10153 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q03701 | |
| CEBPZ | |
| A synthesized peptide derived from human CEBPZ(Accession Q03701), corresponding to amino acid residues V4-D54. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion