Learn More
C/EBP alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA587519
2616.79 DKK valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Description
Immunogen sequence: APAGPGGAVM PGGAHGPPPG YGCAAAGYLD GRLEPLYERV GAPALRPLVI KQEPREEDEA KQLALAGLFP Positive Samples: mouse lung; Cellular Location: Nucleus, Nucleus, nucleolus
The C/EBP (CCAAT/enhancer binding protein) family of transcription factors--alpha, beta, delta, and epsilon)--are widely expressed proteins found in a variety of tissues. These domains contain an evolutionarily conserved five-amino acid motif (the regulatory domain motif RDM) that conforms to the consensus sequence (I/V/L) KXEP. One of the major roles of these transcription factors is that of metabolic regulation in liver and fat cells. Studies suggest that these transcription factors act in a cascade aiding in the development of the adipose cell. Three C/EBP beta isoforms (LAP*, LAP, and LIP) arise from different translation initiation and display different function in gene regulation. From a neurological standpoint, C/EBPs have been implicated in the mediation of nerve growth factor, altering synaptic plasticity, and mediation of inflammatory response in AD patients.
Specifications
C/EBP alpha | |
Polyclonal | |
Unconjugated | |
CEBPA | |
C EBP, C/EBP, c/EBP alpha, C/EBPa, C/ebpalpha, C/EBP-alpha, CAAT/enhancer-binding protein DNA-binding protein, CAAT/enhancer-binding protein, DNA-binding protein, CBF-A, CCAAT enhancer binding protein alpha, CCAAT/enhancer binding protein (C/EBP), alpha, CCAAT/enhancer binding protein alpha, CCAAT/enhancer binding protein, alpha, CCAAT/enhancer-binding protein alpha, Cebp, Cebpa, DBPCEP, OTTHUMP00000220269 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1050, 12606, 24252 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.76 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
P05554, P49715, P53566 | |
CEBPA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 111-180 of human CEBPA (NP_0043552) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.