Få mere at vide
C 87, MedChemExpress
Artikelnummer. 16492636
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16492636 25 mg
16449696 50 mg
16445877 5 mg
16434436 100 mg
16403148 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16492636 Leverandør Medchem Express Leverandørnr. HY100735
MedChemExpress C 87 is a novel small-molecule TNFα inhibitor; potently inhibits TNFα-induced cytotoxicity with an IC50 of 8.73 μM.
C 87 is a novel small-molecule TNFα inhibitor
Tekniske data
Kemisk navn eller materiale C 87
Fysisk form Solid
CAS 332420-90-3
Farve Orange
Mængde 25 mg
Renhedsgrad noter Research
Molekylær formel C24H15ClN6O3S
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : 5.05 mg/mL (10.04 mM; Need ultrasonic)
SMIL O=C(N(C1=NC(C2=CC=CC=C2)=CS1)N=C/3C4=CC=CC=C4)C3=N\NC5=CC=C(Cl)C([N+]([O-])=O)=C5
Molekylvægt (g/mol) 502.93
Formel vægt 502.93
Procent renhed 98.03%