Få mere at vide
Tocris Bioscience™ C 87
TNF-α inhibitor
Brand:  Tocris Bioscience™ 5484/10
Encompass
Produktkode 15726876
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
C 87 is a TNF-α inhibitor (Kd = 110 nM for hTNF-α). Inhibits TNF-α-induced cytotoxicity in L929 fibroblast cells. Attenuates inflammation and increases survival in a mouse hepatitis model.
Tekniske data
Tekniske data
C 87
3-Phenyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(4-chloro-3-nitrophenyl)hydrazone]
Soluble to 20mM in DMSO
Bioactive Small Molecule
332420-90-3
10 mg
Solid
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
Research Use Only. Not For Human or Veterinary Use