Få mere at vide
Tocris Bioscience™ Bz 423
Beskrivelse
Bz 423 is an ATP synthase inhibitor (IC50 ~5 μM). Binds the oligomycin sensitivity conferring protein (OSCP) component of mitochondrial F1F0-ATPase in vitro resulting in generation of superoxide and apoptosis. Kills primary B and T cell in mice in vivo. Suppresses autoimmunity and prolongs survival in mouse lupus model.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 7-Chloro-1,3-dihydro-5-(4-hydroxyphenyl)-1-methyl-3-(2-naphthalenylmethyl)-2H-1,4-benzodiazepin-2-one |
| CAS | 216691-95-1 |
| Mængde | 10 mg |
| Opløselighed | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molekylær formel | C27H21ClN2O2 |
| Formel vægt | 440.92 |
| Fysisk form | Solid |
| Farve | White |
| Produkttype | Bioactive Small Molecule |
| Renhed | >98% |
For Research Use Only.