Få mere at vide
Tocris Bioscience™ BX 430
Selective P2X4 allosteric antagonist
Brand: Tocris Bioscience™ 5545/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BX 430 is a selective P2X4 allosteric antagonist (IC50 = 0.54 μM). Exhibits >10-fold selectivity for human P2X4 receptors over P2X1, P2X2, P2X3, P2X5 and P2X7 receptors. Binds noncompetitively at extracellular allosteric site. Effective at human and zebrafish, but not mouse and rat P2X4 receptors.Tekniske data
| N-[2,6-Dibromo-4-(1-methylethyl)phenyl]-N′-(3-pyridinyl)urea | |
| 688309-70-8 | |
| 10 mg |