missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BVES Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99427AF532
Additional Details : Weight : 0.00970kg
Description
BVES Polyclonal specifically detects BVES in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BVES | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
blood vessel epicardial substance, H, HBVES, POP1MGC42413, POPDC1popeye domain containing 1, Popeye domain-containing protein 1, Popeye protein 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse BVES. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
11149 | |
Store at 4°C in the dark. | |
IgG |