Få mere at vide
Butyryl-d7 Chloride, TRC
Artikelnummer. 30329788
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30329788 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329788 Leverandør Toronto Research Chemicals Leverandørnr. TRCB81051250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butyryl-d7 Chloride
CAS 1219805-71-6
Molekylær formel C4 D7 Cl O
Mængde 50 mg
Synonym Butanoyl-2,2,3,3,4,4,4-d7 chloride (ACI), 2,2,3,3,4,4,4-Heptadeuteriobutanoyl chloride, Butyryl-d7 Chloride, Butanoyl Chloride-D7, Butanoyl chloride-d7
InChI formel InChI=1S/C4H7ClO/c1-2-3-4(5)6/h2-3H2,1H3/i1D3,2D2,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)Cl
IUPAC navn 2,2,3,3,4,4,4-heptadeuteriobutanoyl chloride
Molekylvægt (g/mol) 113.594
Formel vægt 113.062