Få mere at vide
Butyryl-d7 Chloride, CDN
Artikelnummer. 30321106
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321106 0.5 g 0.5g
30317249 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321106 Leverandør CDN Leverandørnr. CDND57790.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butyryl-d7 Chloride
CAS 1219805-71-6
Molekylær formel C4 D7 Cl O
Mængde 0.5 g
Synonym Butanoyl-2,2,3,3,4,4,4-d7 chloride (ACI), 2,2,3,3,4,4,4-Heptadeuteriobutanoyl chloride, Butyryl-d7 Chloride, Butanoyl Chloride-D7, Butanoyl chloride-d7
InChI formel InChI=1 S/C4H7ClO/c1-2-3-4(5)6/h2-3H2,1H3/i1D3,2D2,3D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)Cl
IUPAC navn 2,2,3,3,4,4,4-heptadeuteriobutanoyl chloride
Molekylvægt (g/mol) 113.594
Formel vægt 113.062 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity