Få mere at vide
Butyryl-2,2-d2 Chloride, CDN
Artikelnummer. 30311481
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30311481 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311481 Leverandør CDN Leverandørnr. CDND03501G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanoyl Chloride-2,2-D2
CAS 90587-10-3
Molekylær formel C4 D2 H5 Cl O
Mængde 1 g
Synonym Butyryl-2,2-d2 chloride (7 CI), Butanoyl-2,2-d2 chloride, 2,2-Dideuteriobutanoyl chloride, Butyryl-α,α-d2 Chloride, Butanoyl Chloride-2,2-D2
InChI formel InChI=1 S/C4H7ClO/c1-2-3-4(5)6/h2-3H2,1H3/i3D2
SMIL [2 H]C([2 H])(CC)C(=O)Cl
IUPAC navn 2,2-dideuteriobutanoyl chloride
Molekylvægt (g/mol) 108.5631
Formel vægt 108.0311 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity