Få mere at vide
Butyrophenone-2',3',4',5',6'-d5, CDN
Artikelnummer. 30321150
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321150 1 g -
30315991 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321150 Leverandør CDN Leverandørnr. CDND57761G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butyrophenone-2',3',4',5',6'-D5
CAS 39058-44-1
Molekylær formel C10 D5 H7 O
Mængde 1 g
Synonym 1-Butanone, 1-(phenyl-d5)- (9 CI), Butyrophenone-2',3',4',5',6'-d5, 1-(Phenyl-d5)1-butanone, Butyrophenone-2',3',4',5',6'-D5, Butyrophenone-d5
InChI formel InChI=1 S/C10H12O/c1-2-6-10(11)9-7-4-3-5-8-9/h3-5,7-8 H,2,6H2,1H3/i3D,4 D,5 D,7 D,8 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)CCC)c([2 H])c1[2 H]
IUPAC navn 1-(2,3,4,5,6-pentadeuteriophenyl)butan-1-one
Molekylvægt (g/mol) 153.2325
Formel vægt 153.1202 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity