Få mere at vide
Butyric-4,4,4-d3 Acid, TRC
Artikelnummer. 30331980
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30331980 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331980 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69224150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanoic Acid 4,4,4-D3
CAS 36789-14-7
Molekylær formel C4H5D3O2
Mængde 50 mg
Synonym Butanoic-4,4,4-d3 acid (9CI), Butyric-4,4,4-d3 acid, Butanoic acid 4,4,4-D3
InChI formel InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3
SMIL OC(CCC([2H])([2H])[2H])=O
IUPAC navn butanoic-4,4,4-d3 acid
Molekylvægt (g/mol) 91.12
Formel vægt 91.07