Få mere at vide
Butyric-3,3-d2 Acid, TRC
Artikelnummer. 30331968
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30331968 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331968 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69223925MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanoic Acid 3,3-D2
CAS 64833-96-1
Molekylær formel C4H6D2O2
Mængde 25 mg
Synonym Butanoic-3,3-d2 acid (9CI), Butyric-3,3-d2 acid, Butanoic acid 3,3-D2
InChI formel InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i2D2
SMIL CC([2H])([2H])CC(O)=O
IUPAC navn butanoic-3,3-d2 acid
Molekylvægt (g/mol) 90.12
Formel vægt 90.06