Få mere at vide
Butyric-3,3-d2 Acid, CDN
Artikelnummer. 30319587
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319587 0.1 g 0.1g
30318564 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319587 Leverandør CDN Leverandørnr. CDND12030.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanoic Acid 3,3-D2
CAS 64833-96-1
Molekylær formel C4H6D2O2
Mængde 0.1 g
Synonym Butanoic-3,3-d2 acid (9 CI), Butyric-3,3-d2 acid, Butanoic acid 3,3-D2
InChI formel InChI=1 S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i2D2
SMIL CC([2 H])([2 H])CC(O)=O
IUPAC navn butanoic-3,3-d2 acid
Molekylvægt (g/mol) 90.12
Formel vægt 90.06 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity