Få mere at vide
Butyric-3,3,4,4,4-d5 Acid, TRC
Artikelnummer. 30344795
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30344795 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30344795 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69224650MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanoic Acid 3,3,4,4,4-D5
CAS 156779-02-1
Molekylær formel C4H3D5O2
Mængde 50 mg
Synonym Butanoic-3,3,4,4,4-d5 acid (9CI), Butyric-d5 acid, Butanoic acid 3,3,4,4,4-D5
InChI formel InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3,2D2
SMIL O=C(CC([2H])([2H])C([2H])([2H])[2H])O
IUPAC navn butanoic-3,3,4,4,4-d5 acid
Molekylvægt (g/mol) 93.14
Formel vægt 93.08