Få mere at vide
Butyric-3,3,4,4,4-d5 Acid, CDN
Artikelnummer. 30311515
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30311515 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311515 Leverandør CDN Leverandørnr. CDND52861G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanoic Acid 3,3,4,4,4-D5
CAS 156779-02-1
Molekylær formel C4H3D5O2
Mængde 1 g
Synonym Butanoic-3,3,4,4,4-d5 acid (9 CI), Butyric-d5 acid, Butanoic acid 3,3,4,4,4-D5
InChI formel InChI=1 S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3,2D2
SMIL O=C(CC([2 H])([2 H])C([2 H])([2 H])[2 H])O
IUPAC navn butanoic-3,3,4,4,4-d5 acid
Molekylvægt (g/mol) 93.14
Formel vægt 93.08 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity