Få mere at vide
Butyric-2,2-d2 Acid, TRC
Artikelnummer. 30330745
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30330745 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30330745 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69223650MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butyric-2,2-d2 Acid
CAS 19136-92-6
Molekylær formel C4 D2 H6 O2
Mængde 50 mg
Synonym Butanoic-2,2-d2 acid (9CI), Butyric-2,2-d2 acid (8CI), Butanoic acid 2,2-D2
InChI formel InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i3D2
SMIL [2H]C([2H])(CC)C(=O)O
IUPAC navn 2,2-dideuteriobutanoic acid
Molekylvægt (g/mol) 90.117
Formel vægt 90.065