Få mere at vide
Butyric-2,2-d2 Acid, CDN
Artikelnummer. 30326585
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326585 1 g
30323405 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326585 Leverandør CDN Leverandørnr. CDND09761G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butyric-2,2-d2 Acid
CAS 19136-92-6
Molekylær formel C4 D2 H6 O2
Mængde 1 g
Synonym Butanoic-2,2-d2 acid (9 CI), Butyric-2,2-d2 acid (8 CI), Butanoic acid 2,2-D2
InChI formel InChI=1 S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i3D2
SMIL [2 H]C([2 H])(CC)C(=O)O
IUPAC navn 2,2-dideuteriobutanoic acid
Molekylvægt (g/mol) 90.117
Formel vægt 90.065 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity