Få mere at vide
Butyric-2,2,3,3-d4 Acid, TRC
Artikelnummer. 30344090
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30344090 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30344090 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6922425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanoic Acid 2,2,3,3-D4
CAS 1219794-99-6
Molekylær formel C4H4D4O2
Mængde 5 mg
Synonym Butanoic-2,2,3,3-d4 acid, Butyric-2,2,3,3-d4 acid, Butanoic acid 2,2,3,3-D4
InChI formel InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i2D2,3D2
SMIL CC([2H])(C([2H])(C(O)=O)[2H])[2H]
IUPAC navn butanoic-2,2,3,3-d4 acid
Molekylvægt (g/mol) 92.13
Formel vægt 92.08