missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Butyramide, 98%

Brand:  Alfa Aesar™ A12494.36

Additional Details : CAS Number : 541-35-5 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 11494124

  • 1570.00 DKK / 500g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
Butyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.

Solubility
Soluble in water (170 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

541-35-5
87.122
DNSISZSEWVHGLH-UHFFFAOYSA-N
10927
butanamide
C4H9NO
MFCD00041894
butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid
CHEBI:50724
CCCC(=O)N

Specifications

Butyramide
216°C
500g
C4H9NO
1361528
butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid
DNSISZSEWVHGLH-UHFFFAOYSA-N
butanamide
10927
87.12
1.032
113°C to 116°C
541-35-5
MFCD00041894
14,1592
Soluble in water (170g/L).
CCCC(=O)N
87.122
CHEBI:50724
98%
Videos
SDS Documents