missing translation for 'onlineSavingsMsg'
Learn More

Butyl Methanesulfonate, TRC

Product Code. 30008959
Change view
Click to view available options
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30008959 5 g
30008601 1 g
30010232 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008959 Supplier Toronto Research Chemicals Supplier No. TRCB6940955G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1912-32-9
Molekylær formel C5 H12 O3 S
Molekylvægt (g/mol) 152.21
Synonym Methanesulfonic acid butyl ester, Butyl mesylate, Butyl methanesulfonate, Methanesulfonic acid n-butyl ester, NSC 36060, n-Butyl methanesulfonate
IUPAC navn butyl methanesulfonate
SMIL CCCCOS(=O)(=O)C

Specifications

Kemisk navn eller materiale Butyl methanesulfonate
Renhedsgrad noter GC
Mængde 5 g
InChI formel InChI=1S/C5H12O3S/c1-3-4-5-8-9(2,6)7/h3-5H2,1-2H3
Formel vægt 152.0507
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.