Få mere at vide
Butibufen-d5, TRC
Artikelnummer. 30347701
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30347701 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347701 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68996210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butibufen-d5
CAS 1346603-83-5
Molekylær formel C14H15D5O2
Mængde 10 mg
Synonym alpha-(Ethyl-d5)-4-(2-methylpropyl)benzeneacetic Acid, 2-(4-Isobutylphenyl)butyric Acid-d5, DF 1903Y-d5, alpha-(Ethyl-d5)-4-(2-methylpropyl)benzeneacetic Acid
InChI formel InChI=1S/C14H20O2/c1-4-13(14(15)16)12-7-5-11(6-8-12)9-10(2)3/h5-8,10,13H,4,9H2,1-3H3,(H,15,16)/i1D3,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C(C(=O)O)c1ccc(CC(C)C)cc1
IUPAC navn 3,3,4,4,4-pentadeuterio-2-[4-(2-methylpropyl)phenyl]butanoic acid
Molekylvægt (g/mol) 225.34
Formel vægt 225.1777