Få mere at vide
Butethamine Hydrochloride, LoGiCal
Artikelnummer. 30459212
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30459212 5 mg
1 options
Artikelnummer. 30459212 Leverandør LoGiCal Leverandørnr. LGCFOR1848.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butethamine Hydrochloride
CAS 553-68-4
Analyt- eller komponentnavne Butethamine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C13 H20 N2 O2 . Cl H
Certificeringer/compliance ISO 9001
Synonym Ethanol, 2-[(2-methylpropyl)amino]-, 1-(4-aminobenzoate), hydrochloride (1:1), Butethamine Hydrochloride, Butethamine hydrochloride (6CI), Ethanol, 2-(isobutylamino)-, p-aminobenzoate (ester), monohydrochloride (8CI), Ethanol, 2-[(2-methylpropyl)amino]-, 4-aminobenzoate (ester), monohydrochloride (9CI), 2-(Isobutylamino)ethyl p-aminobenzoate hydrochloride, Ibylcaine, Ibylcaine hydrochloride, Monocaine, Monocaine hydrochloride
InChI formel InChI=1S/C13H20N2O2.ClH/c1-10(2)9-15-7-8-17-13(16)11-3-5-12(14)6-4-11;/h3-6,10,15H,7-9,14H2,1-2H3;1H
Løsningstype Neat
SMIL Cl.CC(C)CNCCOC(=O)c1ccc(N)cc1
IUPAC navn 2-(2-methylpropylamino)ethyl 4-aminobenzoate;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 272.77
Mængde 5 mg
Formel vægt 272.1292 g/mol