Få mere at vide
Butetamate Citrate, LoGiCal
Artikelnummer. 30463723
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463723 10 mg
1 options
Artikelnummer. 30463723 Leverandør LoGiCal Leverandørnr. LGCFOR0606.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butetamate Citrate
CAS 13900-12-4
Analyt- eller komponentnavne Butetamate Citrate
Forsendelsestilstand Room Temperature
Molekylær formel C16 H25 N O2 . C6 H8 O7
Certificeringer/compliance ISO 9001
Synonym Butetamate Citrate, 2-Phenylbutyric acid 2-(diethylamino)ethyl ester citrate, 2-Diethylaminoethyl 2-phenylbutyrate citrate, Butethamate citrate, alpha-Ethylbenzeneacetic acid 2-(diethylamino)ethyl ester 2-hydroxy-1,2,3-propanetricarboxylate
InChI formel InChI=1S/C16H25NO2.C6H8O7/c1-4-15(14-10-8-7-9-11-14)16(18)19-13-12-17(5-2)6-3;7-3(8)1-6(13,5(11)12)2-4(9)10/h7-11,15H,4-6,12-13H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Løsningstype Neat
SMIL CCC(C(=O)OCCN(CC)CC)c1ccccc1.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC navn 2-(diethylamino)ethyl 2-phenylbutanoate;2-hydroxypropane-1,2,3-tricarboxylic acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 455.5
Mængde 10 mg
Formel vægt 455.2155 g/mol