missing translation for 'onlineSavingsMsg'
Få mere at vide

Butenafin hydrochlorid, TRC

Product Code. 30339123
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30339123 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339123 Supplier Toronto Research Chemicals Supplier No. TRCB690195100MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Butenafine Hydrochloride
CAS 101827-46-7
Renhedsgrad noter HPLC
Molekylær formel C23 H27 N . Cl H
Mængde 100 mg
Synonym 1-Naphthalenemethanamine, N-[[4-(1,1-dimethylethyl)phenyl]methyl]-N-methyl-, hydrochloride (1:1), 1-Naphthalenemethanamine, N-[[4-(1,1-dimethylethyl)phenyl]methyl]-N-methyl-, hydrochloride (9CI), Butenafine hydrochloride, KP 363, Mentax, Scorba
InChI formel InChI=1S/C23H27N.ClH/c1-23(2,3)21-14-12-18(13-15-21)16-24(4)17-20-10-7-9-19-8-5-6-11-22(19)20;/h5-15H,16-17H2,1-4H3;1H
SMIL Cl.CN(Cc1ccc(cc1)C(C)(C)C)Cc2cccc3ccccc23
IUPAC navn 1-(4-tert-butylphenyl)-N-methyl-N-(nafthalen-1-ylmethyl)methanamin; hydrochlorid
Molekylvægt (g/mol) 353.93
Formel vægt 353.191
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.