missing translation for 'onlineSavingsMsg'
Få mere at vide

Butanoic-d7 Acid, TRC

Artikelnummer. 30341444
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30341444 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341444 Leverandør Toronto Research Chemicals Leverandørnr. TRCB692237500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Butanoic-d7 Acid
CAS 73607-83-7
Renhedsgrad noter GC
Molekylær formel C4 D7 H O2
Mængde 500 mg
Synonym Butyric-d7 Acid, Butanoic-d7 acid (9CI), Butanoic acid D7
InChI formel InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3,2D2,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,4-heptadeuteriobutanoic acid
Molekylvægt (g/mol) 95.15
Formel vægt 95.0964
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.