Få mere at vide
Butanoic-d7 Acid, TRC
Artikelnummer. 30342234
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30342234 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342234 Leverandør Toronto Research Chemicals Leverandørnr. TRCB692237100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanoic-d7 Acid
CAS 73607-83-7
Renhedsgrad noter GC
Molekylær formel C4 D7 H O2
Mængde 100 mg
Synonym Butyric-d7 Acid, Butanoic-d7 acid (9CI), Butanoic acid D7
InChI formel InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3,2D2,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,4-heptadeuteriobutanoic acid
Molekylvægt (g/mol) 95.15
Formel vægt 95.0964
Procent renhed >95