Få mere at vide
Butanal-d8, TRC
Artikelnummer. 30346532
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30346532 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30346532 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68989210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butanal-d8
CAS 84965-36-6
Molekylær formel C4 2H8 O
Mængde 10 mg
Synonym Butanal-d8, Butyraldehyde-d8, Butal-d8, Butaldehyde-d8, Butanaldehyde-d8, Butyral-d8, Butyric Aldehyde-d8, Butyrylaldehyde-d8, NSC 62779-d8, n-Butanal-d8, n-Butyl Aldehyde-d8, n-Butyraldehyde-d8, n-Butyric Aldehyde-d8
InChI formel InChI=1S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3,2D2,3D2,4D
SMIL [2H]C(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,2,2,3,3,4,4,4-octadeuteriobutan-1-one
Molekylvægt (g/mol) 80.16
Formel vægt 80.1077