Få mere at vide
Butamirate-d5 Citrate, TRC
Artikelnummer. 30339970
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30339970 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339970 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69002710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butamirate-d5 Citrate
CAS 1215650-08-0
Molekylær formel C24H32D5NO10
Mængde 10 mg
Synonym 2-(2-Diethylaminoethoxy)ethyl 3,3,4,4,4-pentadeuterio-2-phenylbutanoate 2-hydroxypropane-1,2,3-tricarboxylic acid, Butamirate-d5 citrate, Butamirate-D5 (C-ethyl-D5) citrate, 2-(2-Diethylaminoethoxy)ethyl 2-phenylbutanoate-3,3,4,4,4-d5 (ester) citrate (salt)
InChI formel InChI=1S/C18H29NO3.C6H8O7/c1-4-17(16-10-8-7-9-11-16)18(20)22-15-14-21-13-12-19(5-2)6-3;7-3(8)1-6(13,5(11)12)2-4(9)10/h7-11,17H,4-6,12-15H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/i1D3,4D2;
SMIL [2H]C([2H])([2H])C([2H])([2H])C(C(=O)OCCOCCN(CC)CC)c1ccccc1.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC navn 2-[2-(diethylamino)ethoxy]ethyl 3,3,4,4,4-pentadeuterio-2-phenylbutanoate;2-hydroxypropane-1,2,3-tricarboxylic acid
Molekylvægt (g/mol) 504.58