missing translation for 'onlineSavingsMsg'
Learn More

Butachlor-d13 (Major), TRC

Product Code. 30341158
Change view
Click to view available options
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30341158 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30341158 Supplier Toronto Research Chemicals Supplier No. TRCB6899271MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Butachlor-d13
CAS 1632119-31-3
Molekylær formel C17H13D13ClNO2
Mængde 1 mg
Synonym N-(Butoxymethyl)-2-chloro-N-(2,6-diethylphenyl)acetamide-d13, 2-Chloro-2',6'-diethyl-N-(butoxymethyl)acetanilide-d13, NSC 221683-d13, 2',6'-Diethyl-N-butoxymethyl-2-chloroacetanilide-d13, Bilchlor-d13, Butaclor-d13, CP 53619-d13, N-(Butoxymethyl)-2-chloro-2',6'-diethylacetanilide-d13, Delchlor 5G-d13, Hiltachlor-d13, Machet-d13, Machete (herbicide)-d13
InChI formel InChI=1S/C17H26ClNO2/c1-4-7-11-21-13-19(16(20)12-18)17-14(5-2)9-8-10-15(17)6-3/h8-10H,4-7,11-13H2,1-3H3/i2D3,3D3,5D2,6D2,8D,9D,10D
SMIL O=C(CCl)N(COCCCC)C1=C(C([2H])([2H])C([2H])([2H])[2H])C([2H])=C([2H])C([2H])=C1C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn N-(butoxymethyl)-2-chloro-N-[3,4,5-trideuterio-2,6-bis(1,1,2,2,2-pentadeuterioethyl)phenyl]acetamide
Molekylvægt (g/mol) 324.93
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.