Få mere at vide
Butachlor-d13 (Major), TRC
Artikelnummer. 30337084
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30337084 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337084 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68992710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butachlor-d13
CAS 1632119-31-3
Molekylær formel C17H13D13ClNO2
Mængde 10 mg
Synonym N-(Butoxymethyl)-2-chloro-N-(2,6-diethylphenyl)acetamide-d13, 2-Chloro-2',6'-diethyl-N-(butoxymethyl)acetanilide-d13, NSC 221683-d13, 2',6'-Diethyl-N-butoxymethyl-2-chloroacetanilide-d13, Bilchlor-d13, Butaclor-d13, CP 53619-d13, N-(Butoxymethyl)-2-chloro-2',6'-diethylacetanilide-d13, Delchlor 5G-d13, Hiltachlor-d13, Machet-d13, Machete (herbicide)-d13
InChI formel InChI=1S/C17H26ClNO2/c1-4-7-11-21-13-19(16(20)12-18)17-14(5-2)9-8-10-15(17)6-3/h8-10H,4-7,11-13H2,1-3H3/i2D3,3D3,5D2,6D2,8D,9D,10D
SMIL O=C(CCl)N(COCCCC)C1=C(C([2H])([2H])C([2H])([2H])[2H])C([2H])=C([2H])C([2H])=C1C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn N-(butoxymethyl)-2-chloro-N-[3,4,5-trideuterio-2,6-bis(1,1,2,2,2-pentadeuterioethyl)phenyl]acetamide
Molekylvægt (g/mol) 324.93